Order Entry
United Kingdom
ContactUsLinkComponent
Anti-MCAK Polyclonal Antibody
Anti-MCAK Polyclonal Antibody
Catalog # BSBTPB9230
Supplier:  Boster Bio
undefined
Anti-MCAK Polyclonal Antibody
Catalog # BSBTPB9230
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    MCAK
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Polyclonal antibody for MCAK/KIF2C detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. MCAK/KIF2C information: Molecular Weight: 81313 MW; Subcellular Localization: Cytoplasm, cytoskeleton . Nucleus . Chromosome, centromere. Chromosome, centromere, kinetochore. Associates with the microtubule network at the growing distal tip (the plus-end) of microtubules, probably through interaction with MTUS2/TIP150 and MAPRE1 (By similarity). Association with microtubule plus ends is also mediated by interaction with KIF18B. Centromeric localization requires the presence of BUB1 and SGOL2; Tissue Specificity: Expressed at high levels in thymus and testis, at low levels in small intestine, the mucosal lining of colon, and placenta, and at very low levels in spleen and ovary; expression is not detected in prostate, peripheral blood Leukocytes, heart, brain, lung, liver, skeletal muscle, kidney or pancreas. Isoform 2 is testis-specific.

Type: Primary
Antigen: MCAK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat