Roxithromycin 96%
ACRO459570050
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Water:≤3 %
- HPLC:≥95.0 %
- Residue after ignition:≤0.1 %
:
:837.04 g/mol
:111…118 °C
:>110 °C
:GHS07
:80214-83-1
:C₄₁H₇₆N₂O₁₅
:MFCD00214389