1,2-Indanedione 95%
Catalog # ACRO460920010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to yellow Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:110 to 120 °C
- Water:≤2.5 %
- HPLC:≥94 %
Specification Test Results
About this item
Synonyms: 1H-Indene-1,2(3H)-dione, 1,2-Indandione
:
:146.15 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:16214-27-0
Formula:C₉H₆O₂
MDL Number:MFCD00463378
Melting Pt:117…124 °C