4-Chloro-1H-imidazole 98%
Catalog # ACRO460030010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to off-white Solid
- Infrared spectrum:Conforms
- Melting point:118 to 121 °C
- Titration with HClO4:≥97.5 %
Specification Test Results
About this item
:
Melting Pt:118…120 °C
:102.52 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:15965-31-8
Formula:C₃H₃ClN₂
MDL Number:MFCD09033087