4-Nitrophenyl phosphate 97%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 4-Nitrophenyl dihydrogen phosphate, pNpp, p-Nitrophenyl phosphate
- CAS Number:330-13-2
- Formula:C₆H₆NO₆P
- Molecular Weight:219.09 g/mol
- Boiling Pt:458 °C
- Melting Pt:153 °C
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
Synonyms: 4-Nitrophenyl dihydrogen phosphate, pNpp, p-Nitrophenyl phosphate
- CAS Number:330-13-2
- Formula:C₆H₆NO₆P
- Molecular Weight:219.09 g/mol
- Boiling Pt:458 °C
- Melting Pt:153 °C
- Storage Temperature:Ambient
Specifications for Product Family
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



