Norbornene 99% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:498-66-8
:
Formula:C₇H₁₀
MDL Number:MFCD00082304
:94.16 g/mol
Boiling Pt:96 °C (760 mmHg)
Melting Pt:42…46 °C
Flash Pt:–15 °C
UN:1325
ADR:4.1,II
:GHS02
:GHS07
:GHS08
:GHS09
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White Crystalline mass
- Infrared spectrum:Conforms
- Melting point:42 to 46 °C
- GC:≥98.5 %
- Stabilizer:0.1 % phenolic antioxidant
- Color scale:≤15 APHA
- Residue after evaporation:≤0.002 %
- Chloride (Cl):≤10 ppm
- Sulfate (SO4):≤10 ppm
- Cadmium (Cd):≤0.05 ppm
- Calcium (Ca):≤0.5 ppm
- Chromium (Cr):≤0.02 ppm
- Cobalt (Co):≤0.02 ppm
- Copper (Cu):≤0.02 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.1 ppm
- Magnesium (Mg):≤0.1 ppm
- Manganese (Mn):≤0.02 ppm
- Nickel (Ni):≤0.02 ppm
- Zinc (Zn):≤0.1 ppm
- Heavy metals (as Pb):≤1 ppm
Specification Test Results
Frequently Bought Together