Order Entry
United Kingdom
ContactUsLinkComponent
Bovine mucin (from Submaxillary glands)
Bovine mucin (from Submaxillary glands)
Catalog # 499643-500
CAS Number:  
Bovine mucin (from Submaxillary glands)
Catalog # 499643-500
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    500 mg
  • Source:
    Submaxillary glands
  • Species:
    Bovine
  • Storage conditions:
    +2...+8 °C
  • Protein/peptide name:
    Mucin
  • Shipping temperature:
    Ambient

Specifications

About this item

Native mucin from bovine submaxillary gland is a high molecular weight glycoprotein and its major constituent of saliva, gastrointestinal juices and other secretions.

  • Lyophilised form
  • Hygroscopic
  • Standard handling

Mucin exists as an aggregated complex in excess of 106 molecular weight. It is a rich source of O-linked oligosaccharide chains and it is suitable as a substrate for neuraminidase.