Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Desloratadine, Reference Standards

 :   USP
Desloratadine, Reference Standards
 :  USP
 
Desloratadine, Reference Standards

 

 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

 

 

 

  • Catalog No:
  • 1173042.
  • 1173053.
  • 1173086.
  • 1173064.
  • Description:
  • Desloratadine
  • Desloratadine Related Compound A
  • Desloratadine Related Compound B
  • Desloratadine
  • Synonym(s):
  • 8-Bromo-6,11-dihydro-11-(piperidin-4-ylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine
  • 8-chloro-11-(1,2,3,6-tetrahydropyridin-4-yl)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine hydrochloride
  • 8-Chloro-6,11-dihydro-11-(N-formyl-4-piperidinylidene) -5H-benzo[5,6]cyclohepta[1,2-b]pyridine
  • CAS No.:
  • 100643-71-8
  • 117796-50-6
  • 183198-49-4
  • 117810-61-4
  • Formula:
  • C₁₉H₁₉ClN₂
  • C₁₉H₁₉BrN₂
  • C₁₉H₁₉ClN₂
  • C₂₀H₁₉ClN₂O

 

Product Family Options

 

  • Description
    Standards
    Pack type
    Pk
     
     
  • Desloratadine
    USP reference standard
    Vial
    200 mg
       
       
    1 * 200 mg
    1 * 200 mg - £388.00
    £388.00
    1 * 200 mg
       £388.00

     
     
    Specifications
    Cat. No.
    1173042.
    Description
    Desloratadine
    Synonym(s)
    CAS No.
    100643-71-8
    Formula
    C₁₉H₁₉ClN₂
  • Desloratadine Related Compound A
    USP reference standard
    Vial
    20 mg
       
       
    1 * 20 mg
    1 * 20 mg - £1,180.00
    £1,180.00
    1 * 20 mg
       £1,180.00

     
     
    Specifications
    Cat. No.
    1173053.
    Description
    Desloratadine Related Compound A
    Synonym(s)
    8-Bromo-6,11-dihydro-11-(piperidin-4-ylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine
    CAS No.
    117796-50-6
    Formula
    C₁₉H₁₉BrN₂
  • Desloratadine Related Compound B
    USP reference standard
    Vial
    20 mg
       
       
    1 * 20 mg
    1 * 20 mg - £1,300.00
    £1,300.00
    1 * 20 mg
       £1,300.00

     
     
    Specifications
    Cat. No.
    1173086.
    Description
    Desloratadine Related Compound B
    Synonym(s)
    8-chloro-11-(1,2,3,6-tetrahydropyridin-4-yl)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine hydrochloride
    CAS No.
    183198-49-4
    Formula
    C₁₉H₁₉ClN₂
  • Desloratadine
    USP reference standard
    Vial
    50 mg
       
       
    1 * 50 mg
    1 * 50 mg - £1,180.00
    £1,180.00
    1 * 50 mg
       £1,180.00

     
     
    Specifications
    Cat. No.
    1173064.
    Description
    Desloratadine
    Synonym(s)
    8-Chloro-6,11-dihydro-11-(N-formyl-4-piperidinylidene) -5H-benzo[5,6]cyclohepta[1,2-b]pyridine
    CAS No.
    117810-61-4
    Formula
    C₂₀H₁₉ClN₂O