Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Metoclopramide Related Compounds, Reference Standard

Metoclopramide Related Compounds, Reference Standard

Supplier:   USP
Metoclopramide Related Compounds, Reference Standard
Supplier:  USP
Certificates
Metoclopramide Related Compounds, Reference Standard

4 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1440819.
  • 1440820.
  • 1440841.
  • 1440808.
  • Description:
  • Metoclopramide Related Compound A
  • Metoclopramide Related Compound B
  • Metoclopramide Related Compound D
  • Metoclopramide Hydrochloride
  • Synonym(s):
  • 4-Acetamido-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide
  • Methyl 4-acetamido-5-chloro-2-methoxybenzoate
  • Methyl 4-acetamido-2-methoxybenzoate
  • CAS No.:
  • 5608-13-9
  • 4093-31-6
  • 4093-29-2
  • 54143-57-6
  • Formula:
  • C₁₆H₂₄ClN₃O₃
  • C₁₁H₁₂ClNO₄
  • C₁₁H₁₃NO₄
  • C₁₄H₂₅Cl₂N₃O₃

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1440819.
    Metoclopramide Related Compound A
    Metoclopramide Related Compound A
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - £1,140.00
    £1,140.00
    1 * 15 mg
       £1,140.00

    Specifications:
    More
    Specifications
    Cat. No.
    1440819.
    Description
    Metoclopramide Related Compound A
    Synonym(s)
    4-Acetamido-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide
    CAS No.
    5608-13-9
    Formula
    C₁₆H₂₄ClN₃O₃
  • 1440820.
    Metoclopramide Related Compound B
    Metoclopramide Related Compound B
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - £1,150.00
    £1,150.00
    1 * 15 mg
       £1,150.00

    Specifications:
    More
    Specifications
    Cat. No.
    1440820.
    Description
    Metoclopramide Related Compound B
    Synonym(s)
    Methyl 4-acetamido-5-chloro-2-methoxybenzoate
    CAS No.
    4093-31-6
    Formula
    C₁₁H₁₂ClNO₄
  • 1440841.
    Metoclopramide Related Compound D
    Metoclopramide Related Compound D
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - £1,150.00
    £1,150.00
    1 * 15 mg
       £1,150.00

    Specifications:
    More
    Specifications
    Cat. No.
    1440841.
    Description
    Metoclopramide Related Compound D
    Synonym(s)
    Methyl 4-acetamido-2-methoxybenzoate
    CAS No.
    4093-29-2
    Formula
    C₁₁H₁₃NO₄
  • 1440808.
    Metoclopramide Hydrochloride
    Metoclopramide Hydrochloride
    USP reference standard
    Vial
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - £388.00
    £388.00
    1 * 500 mg
       £388.00

    Specifications:
    More
    Specifications
    Cat. No.
    1440808.
    Description
    Metoclopramide Hydrochloride
    Synonym(s)
    CAS No.
    54143-57-6
    Formula
    C₁₄H₂₅Cl₂N₃O₃
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.