Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Iopromide, Reference Standards

Iopromide, Reference Standards

Supplier:   USP
Iopromide, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1344804.
  • 1344826.
  • 1344837.
  • Description:
  • Iopromide
  • Iopromide Related Compound A
  • Iopromide Related Compound B
  • Synonym(s):
  • 5-Amino-N1,N3-Bis-(2,3-dihydroxypropyl)-2,4,6-triiodo-N1-methylisophthalamide
  • 5-(Acetylamino)-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-N-methyl-1,3-benzenedicarboxamide
  • CAS No.:
  • 73334-07-3
  • 154361-51-0
  • 76350-28-2
  • Formula:
  • C₁₈H₂₄I₃N₃O₈
  • C₁₅H₂₀I₃N₃O₆
  • C₁₇H₂₂I₃N₃O₇

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1344804.
    Iopromide
    Iopromide
    USP reference standard
    Vial
    400 mg
    Log in for availability
    Log in for availability
    1 * 400 mg
    1 * 400 mg - £338.00
    £338.00
    1 * 400 mg
       £338.00

    Specifications:
    More
    Specifications
    Cat. No.
    1344804.
    Description
    Iopromide
    Synonym(s)
    CAS No.
    73334-07-3
    Formula
    C₁₈H₂₄I₃N₃O₈
  • 1344826.
    Iopromide Related Compound A
    Iopromide Related Compound A
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - £1,180.00
    £1,180.00
    1 * 50 mg
       £1,180.00

    Specifications:
    More
    Specifications
    Cat. No.
    1344826.
    Description
    Iopromide Related Compound A
    Synonym(s)
    5-Amino-N1,N3-Bis-(2,3-dihydroxypropyl)-2,4,6-triiodo-N1-methylisophthalamide
    CAS No.
    154361-51-0
    Formula
    C₁₅H₂₀I₃N₃O₆
  • 1344837.
    Iopromide Related Compound B
    Iopromide Related Compound B
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - £1,190.00
    £1,190.00
    1 * 50 mg
       £1,190.00

    Specifications:
    More
    Specifications
    Cat. No.
    1344837.
    Description
    Iopromide Related Compound B
    Synonym(s)
    5-(Acetylamino)-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-N-methyl-1,3-benzenedicarboxamide
    CAS No.
    76350-28-2
    Formula
    C₁₇H₂₂I₃N₃O₇
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.