Mouse recombinant CD275 (from CHO cells)
PRSI90-436
:
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Mouse
- Tag sequence:Fc Tag
- Storage conditions:Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
- Protein synonyms:ICOS Ligand|B7-H2|B7RP-1|B7-like Protein Gl50
- Protein/peptide name:CD275
- Purity:>98% (SDS-PAGE)
- Sequence:48-279
- Formulation:Lyophilized from 0.2um-filtered solution in PBS.
Mouse B7-H2, also called CD275, B7RP-1, B7h, LICOS and GL50, is a member of the growing B7 family of immune costimulatory proteins. CD275 has been identified as the ligand for ICOS, a member of the CD28 family of co-stimulatory receptors. Mouse CD275 is expressed on resting B cells and at low levels on monocytes. The CD275/ ICOS interaction appears to play roles in T cell dependent B cell activation and Th differentiation.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.