Order Entry
United Kingdom
ContactUsLinkComponent
Mouse recombinant CD86 (from CHO cells)
Mouse recombinant CD86 (from CHO cells)
Catalog # PRSI90-406
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant CD86 (from CHO cells)
Catalog # PRSI90-406
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Mouse
  • Tag sequence:
    Fc Tag
  • Storage conditions:
    Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Protein synonyms:
    B70|ETC-1|B7-2
  • Protein/peptide name:
    CD86
  • Purity:
    >98% (SDS-PAGE)
  • Sequence:
    26-245
  • Formulation:
    Lyophilized from 0.2um-filtered solution in PBS.

Specifications

About this item

CD86 (B7-2) is a 60-100 kDa variably glycosylated protein in the B7 family. B7 family members are transmembrane cell surface molecules that play important roles in immune activation and the maintenance of immune tolerance. B7-2 is highly expressed on activated antigen presenting cells (APC), e.g. B cells, dendritic cells and monocytes as well as on vascular endothelial cells. B7-2 and the closely related CD80 (B7-1) exhibit overlapping but distinct functional properties. Their binding to CD28, which is constitutively expressed on T cells, enhances T cell receptor signalling and also provides TCR-independent costimulation. B7-1 and B7-2 additionally bind the CD28-related protein CTLA-4, which is up-regulated and recruited to the immunological synapse (IS) at the onset of T cell activation. CTLA-4 ligation inhibits the T cell response and supports regulatory T cell function. B7-2 is expressed earlier than B7-1 following APC activation and both proteins bind with higher affinity to CTLA-4 than to CD28. B7-2 promotes the stabilisation of CD28 in the IS, while B7-1 is primarily responsible for promoting CTLA-4 recruitment and accumulation in the IS. The relative participation of B7-1 and B7-2 in T cell costimulation can also alter the Th1/Th2 bias of the immune response. Both B7-1 and B7-2 serve as cellular receptors for B species adenoviruses.

  • This recombinant proteins is for research use only.

Fusion-Tag: Fc Tag

This recombinant proteins is for research use only.