Order Entry
United Kingdom
ContactUsLinkComponent
 
Human recombinant FSH (from Cells)
Catalog # PRSI92-620
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant FSH (from Cells)
Catalog # PRSI92-620
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    FSH alpha/beta|FSH|Follicle-stimulating hormone
  • Protein/peptide name:
    FSH
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    10.2 and 12.5 kD
  • Sequence:
    Ala25-Ser116&Asn19-Glu129
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

Human Follicle-stimulating hormone (FSH) is a member of glycoprotein hormones subunit beta family, which also includes LH, chorionic gonadotropin (CG) and thyroid-stimulating hormone (TSH). FSH and its family members are heterodimers consisting of non-covalently linked alpha- and beta -subunits. They share an identical alpha subunit, and beta -subunits vary. FSH has a unique beta -subunit (FSH beta ), which confers its specific biologic activity and is responsible for interaction with the FSH-receptor which belongs to a subfamily of GPCRs called leucine-rich-repeat-containing GPCRs (LGRs). FSH is secreted from the pituitary gland and regulates reproduction in mammals. FSH stimulates Sertoli cell proliferation in testes and supports spermatogenesis in males, and induces the maturation of ovarian follicles in females.

This recombinant protein can be used for biological assays. For research use only.