Mouse recombinant TIMP1
Catalog # PRSI92-603
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:Mouse
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:Erythroid-Potentiating Activity|Metalloproteinase Inhibitor 1|Tissue Inhibitor of Metalloproteinases 1|CLGI|TIMP1|TIMP|Collagenase Inhibitor|Fibroblast collagenase Inhibitor|EPA|TIMP-1
- Protein/peptide name:TIMP1
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:20.2 kD
- Sequence:Cys25-Arg205
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM Tris,150mM NaCl,pH8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
Specifications
About this item
Tissue Inhibitor of Metalloproteinases 1 (TIMP-1) complexes with metalloproteinases (such as collagenases) and irreversibly inactivates them by binding to their catalytic zinc cofactor. Also mediates erythropoiesis in vitro; but, unlike IL-3, it is species-specific, stimulating the growth and differentiation of only human and murine erythroid progenitors. Known to act on MMP-1, MMP-2, MMP-3, MMP-7, MMP-8, MMP-9, MMP-10, MMP-11, MMP-12, MMP-13, and MMP-16. TIMP-1 does not act on MMP-14.
This recombinant protein can be used for biological assays. For research use only.