Order Entry
United Kingdom
ContactUsLinkComponent
Human recombinant lysozyme C (from cells)
Human recombinant lysozyme C (from cells)
Catalog # PRSI92-394
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant lysozyme C (from cells)
Catalog # PRSI92-394
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    LZM|Lysozyme C|4-beta-N-acetylmuramidase C|1|LYZ
  • Protein/peptide name:
    Lysozyme C
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    15.7 kD
  • Sequence:
    Lys19-Val148
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl,10% Glycerol,pH7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

lysozyme C is a secreted protein and belongs to the glycosyl hydrolase 22 family. Lysozymes have primarily a bacteriolytic function, damage bacterial cell walls by catalyzing hydrolysis of 1,4-beta-linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues in a peptidoglycan and between N-acetyl-D-glucosamine residues in chitodextrins. Those in tissues and body fluids are associated with the monocyte-macrophage system and enhance the activity of immunoagents. Lysozyme C is capable of both hydrolysis and transglycosylation; it shows also a slight esterase activity. It acts rapidly on both peptide-substituted and unsubstituted peptidoglycan, and slowly on chitin oligosaccharides.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.