Human recombinant serum amyloid A4 (from E. coli)
PRSI92-072
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Serum Amyloid A-4 Protein|Constitutively Expressed Serum Amyloid A Protein|C-SAA|CSAA|SAA4
- Protein/peptide name:Serum Amyloid A4
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:14 kD
- Sequence:Glu19-Thr130
- Formulation:Supplied as a 0.2 um filtered solution of 20mM Tris, 200mM NaCl, 0.5mM EDTA, 20% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Serum Amyloid A-4 Protein (SAA4) is a member of the SAA family. SAA proteins are a family of apolipoproteins associated with high-density lipoprotein (HDL) in plasma. SAA4 is constitutively expressed only in humans and mice. Its physiological function is unknown. SAA4 functions as a major acute phase reactant. SAA4 mRNA and protein occurrence in macrophage derived foam cells of coronary and carotid arteries implied a specific role of human SAA4 during inflammation including atherosclerosis.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag