Human recombinant MBIPP (from E. coli)
Catalog # PRSI92-032
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Environmentally Preferable:
- Tag sequence:N-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:MAPK Upstream Kinase-Binding Inhibitory Protein|MBIP|MUK-Binding Inhibitory Protein|MAP3K12-Binding Inhibitory Protein 1
- Protein/peptide name:MBIPP
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:42.51 kD
- Sequence:Met1-Pro344
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM Tris, pH8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
MAP3K12-binding inhibitory protein 1 (MBIP) is a 39kD protein high expression in the heart and lung. It is a component of the ADA2A-containing complex (ATAC) complex, a complex with histone acetyltransferase activity on histones H3 and H4, and composed of CSRP2BP, KAT2A, TADA2L, TADA3L, ZZ3, MBIP, WDR5, YEATS2, CCDC101 and DR1. In the complex, it probably interacts directly with KAT2A, CSRP2BP and WDR5. It’s function to inhibit the MAP3K12 activity to induce the activation of the JNK/SAPK pathway.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag