Human recombinant IZUMO4 (from cells)
Catalog # PRSI91-963
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-Fc tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:Sperm 22 kDa protein c113|C19orf36|UNQ831/PRO1758|IZUMO4
- Protein/peptide name:IZUMO4
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:50.1 kD
- Sequence:His16-His214
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:used for biological assays
Specifications
About this item
Izumo sperm-egg fusion protein 4 is a sperm membrane protein which plays a key role in the fusion in the mouse. IZUMO4 has an N-terminal domain with significant homology to the N-terminal domain of Izumo.It belongs to the Izumo family . Izumo 4 is a soluble protein expressed in the testis and in other tissues. Izumo domain possesses the ability to form dimers, whereas the transmembrane domain or the cytoplasmic domain or both of Izumo 1 are required for the formation of multimers of higher order.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag