Human recombinant LRG1 (from cells)
Catalog # PRSI91-951
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:HMFT1766|LRG1|Leucine-rich alpha-2-glycoprotein|LRG
- Protein/peptide name:LRG1
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:35.3 kD
- Sequence:Val36-Gln347
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM Tris, 20mM NaCl, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Leucine-rich alpha-2-glycoprotein is a secreted protein and contains 8 LRR (leucine-rich) repeats and 1 LRRCT domain. The leucine-rich repeat (LRR) family of proteins, including LRG1, have been shown to be involved in protein-protein interaction, signal transduction, and cell adhesion and development. LRG1 is expressed during granulocyte differentiation. Levels of the LRG protein are markedly elevated in acute appendicitis and therefore could be used as a diagnostic aid.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag