Human recombinant Clusterin-like 1 (from cells)
Catalog # PRSI91-760
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:CLUL1|Retinal-Specific Clusterin-like Protein|Clusterin-like Protein 1
- Protein/peptide name:Clusterin-like 1
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:50.06 kD
- Sequence:Ala21-Ala442
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM Tris,150mM NaCl,pH8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Clusterin-Like Protein 1 (CLUL1) is a secreted protein that belongs to the clusterin family. CLUL1 is synthesized as a 466 amino acid precursor that contains a 20 amino acid signal sequence, and a 446 amino acid mature chain. CLUL1 is expressed predominantly by cone photoreceptors of the retina. It has been shown that CLUL1 expression is down-regulated in some forms of retinal disease.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag