Recombinant protein L (from E. coli)
Catalog # PRSI96-656
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:2 mg
- Protein/peptide type:Recombinant
- Source:E. coli
- Protein synonyms:RPL|Protein L
- Protein/peptide name:Protein L
- Purity:>98% as determined by SDS-PAGE.
- Formulation:Lyophilized, PBS, pH7.4
- Tested applications:Western Blot