Order Entry
United Kingdom
ContactUsLinkComponent
Human recombinant PDL18 (from HEK293 cells)
Human recombinant PDL18 (from HEK293 cells)
  PRSI96-598
 :  
undefined
Human recombinant PDL18 (from HEK293 cells)
  PRSI96-598
 :  

 

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Biological activity:
    Measured by its binding ability in a functional ELISA. Immobilized Human PD-L1 /B7-H1 /CD274 Protein, C-Fc Tag at 5μg/mL (100 μl/well),can bind Biotinylated Human PDCD1 Protein, C-Fc, His & AVI Tagged. The EC50 is typically 16. 4-196. 8 ng/mL. Measured by its binding ability in a functional ELISA. Immobilized Human PD-L1 /B7-H1 /CD274 Protein, C-Fc Tag at 5μg/mL (100 μl/well),can bind Biotinylated Human PDCD1 Protein,C-Fc Tag. The EC50 is typically 25-227 ng/mL.
  • Protein synonyms:
    B7-H1|PD-L1|CD274|PDCD1L1|PDCD1LG1
  • Protein/peptide name:
    PDL18
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    51.4 kDa
  • Formulation:
    Lyophilized, 50 mM tris, 100 mM glycine, pH7.5
  • Tested applications:
    Western Blot

 

 

Programmed cell death 1 ligand 1 (PD-L1) is also known as cluster of differentiation (CD274) or B7 homolog 1 (B7-H1), is a member of the growing B7 family of immune molecules and is involved in the regulation of cellular and humoral immune responses. B7-H1 is a cell surface immunoglobulin superfamily with two Ig-like domains within the extracellular region and a short cytoplasmic domain. PD-L1 is highly expressed in the heart, skeletal muscle, placenta and lung and weakly expressed in the thymus, spleen, kidney and liver. PD-L1 is expressed on activated T-cells, B-cells, dendritic cells, keratinocytes and monocytes. PD-L1 is up-regulated on T- and B-cells, dendritic cells, keratinocytes and monocytes after LPS and IFNG activation and up-regulated in B-cells activated by surface Ig cross-linking. PD-L1 involve in the costimulatory signal, essential for T-cell proliferation and production of IL10 and IFNG, in an IL2-dependent and a PDCD1-independent manner.