Human recombinant CD3 delta (from HEK293 cells)
PRSI96-160
:
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Environmentally Preferable:
- Protein synonyms:CD3-DELTA|T3D|CD3D
- Protein/peptide name:CD3 delta
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:14.3 kDa
- Formulation:Lyophilized, PBS, pH7.4
- Tested applications:Western Blot