Human recombinant IL1B (fromE. coli)
PRSI40-598
:
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Biological activity:Determined by its ability to stimulate the proliferation of mouse D10S cells. The expected ED50 is = 0.001 ng/mL, corresponding to a specific activity of = 1 x 109 units/mg.
- Protein synonyms:IL-1beta
- Protein/peptide name:IL1B
- Purity:>98%
- Formulation:Lyophilized
IL-1β is a proinflammatory cytokine produced in a variety of cells including monocytes, tissue macrophages, keratinocytes and other epithelial cells. Both IL-1α and IL-1β binds to the same receptor and has similar if not identical biological properties. These cytokines have a broad range of activities including, stimulation of thymocyte proliferation, by inducing IL-2 release, B-cell maturation and proliferation, mitogenic FGF-like activity and the ability to stimulate the release of prostaglandin and collagenase from synovial cells. However, whereas IL-1β is a secreted cytokine, IL-1β is predominantly a cell-associated cytokine. Recombinant human IL-1β is a 17.3 kDa protein containing 153 amino acid residues.
- Store in working aliquots at -20°C. Avoid freeze/thaw cycles.