Human SCUBE3 peptide
Catalog # PRSI5847P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Protein/peptide name:SCUBE3 Peptide
- Concentration:200 ug/mL
- Formulation:Liquid
Specifications
About this item
15 amino acids near the center of human SCUBE3.
- Can be used to block antibody activity completely in WB by incubating with equal volume of antibody for 30 min at 37°C.
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide