Human BCAR3 peptide
Catalog # PRSI6999P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:16 amino acid peptide near the amino terminus of human BCAR3
- Protein/peptide name:BCAR3 Peptide
- Tested applications:Blocking
Specifications
About this item
BCAR3 peptide is used for blocking the activity of BCAR3 antibody.