Order Entry
United Kingdom
ContactUsLinkComponent
Human ARTS peptide
Human ARTS peptide
Catalog # PRSI3025P
Supplier:  ProSci Inc.
CAS Number:  
Human ARTS peptide
Catalog # PRSI3025P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Source:
  • Species:
    Human
  • Environmentally Preferable:
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    12 amino acids terminus of human ARTS Peptide
  • Protein/peptide name:
    ARTS Peptide
  • Tested applications:
    Blocking

Specifications

About this item

ARTS peptide is used for blocking the activity of ARTS antibody.