Anti-FGF19 Mouse Monoclonal Antibody [clone: FG369-1]
Catalog # PRSI36-067
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:fibroblast growth factor 19
- Antigen symbol:FGF19
- Clonality:Monoclonal
- Clone:FG369-1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:9965
- Antigen synonyms:Fibroblast Growth Factor 19
- Storage buffer:0.2um-filtered solution in PBS, pH 7.4. Contains no preservatives.
- Storage temperature:Stable for at least 1 year after receipt when stored at -20˚C.
- Concentration:1 mg/ml
- Shipping temperature:-20°C
- Immunogen:Recombinant human FGF-19.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
FGF-19 is involved in the suppression of bile acid biosynthesis through down-regulation of CYP7A1 expression, following positive regulation of the JNK and ERK1/2 cascades. Stimulates glucose uptake in adipocytes. FGF-19 plays an important role in brain development during embryogenesis. FGF-19 binds only to FGF receptor 4 (FGFR4). Activity requires the presence of KLB.
Type: Primary
Antigen: FGF19
Clonality: Monoclonal
Clone: FG369-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human