Anti-CCL7 Mouse Monoclonal Antibody [clone: PEPY042_hMCP3]
PRSIXP-5733-M
- Antibody type:Primary
- Antigen name:Chemokine (C-C motif) ligand 7
- Antigen symbol:CCL7
- Clonality:Monoclonal
- Clone:PEPY042_hMCP3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Neutralisation:Yes
- Form:Lyophilized
- Gene ID:6354
- Antigen synonyms:MCP3|NC28|SCYA7|SCYA6|MCP-3|MARC|FIC
- Storage temperature:Stable at room temperature for at least one month and for greater than a year when kept at –20 °C. Reconstitute in sterile water to a concentration of 0.1 - 1.0 mg/mL. Stable for at least six weeks at 2-4 °C when reconstituted.
- Shipping temperature:RT
- Immunogen:The monoclonal antibody was produced in C3H/HeJ mice using recombinant hMCP-3 as the immunizing antigen.
- Purification:This IgG1 antibody was purified from ascites fluid on a Protein A affinity column.
- Size:500 µg
- Pk:500 µG
MCP-3 is a secreted chemokine which attracts macrophages during inflammation and metastasis. It is a member of the C-C subfamily of chemokines which are characterized by having two adjacent cysteine residues. The protein is an in vivo substrate of matrix metalloproteinase 2, an enzyme which degrades components of the extracellular matrix.
Type: Primary
Antigen: CCL7
Clonality: Monoclonal
Clone: PEPY042_hMCP3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human