Anti-CXCL12 Rabbit Polyclonal Antibody (Biotin)
Catalog # PRSIXP-5269BT
Supplier: ProSci Inc.
Specifications
- Antigen name:Chemokine (C-X-C Motif) Ligand 12
- Antigen symbol:CXCL12
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:SDF1B|TPAR1|SDF1|IRH|SDF1A|Stromal cell-derived factor 1|PBSF|SDF-1|TLSF|SCYB12
- Immunogen:Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hSDF-1-β (human SDF-1-β).
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Anti-hSDF-1-β specific antibody was purified by affinity chromatography and then biotinylated.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CXCL12
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: