Specifications
- Antigen name:Tumor Necrosis Factor Receptor Superfamily Member 21
- Antigen symbol:TNFRSF21
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Rat
- Isotype:IgG2a
- Reactivity:Human
- Form:Liquid
- Antigen synonyms:Tumor necrosis factor receptor superfamily member 21|Death receptor 6|UNQ437/PRO868|DR6|CD358|BM-018
- Immunogen:DR6 (monoclonal Luke-1) antibody was raised against recombinant human DR6;Fc (aa. 42-335).
- Tested applications:Fluorescence-Activated Cell Sorting
- Size:100 µg
- Pk:100 µG
Specifications
About this item
DR6 antibody can be used to recognize human DR6 by Flow Cytometry. Method: Cells (5x105) were incubated on ice for 30 min in 50 μl FACS buffer (PBS, 5% FCS, 0.02% azide) containing 1 μg/mL of anti-DR6 MAb (Luke-1). After washing in FACS buffer, R-PE-conjugated antibody to rat IgG was added. Cells were incubated on ice for 30 min, washed and analyzed by flow cytometry. Method: 3 HEK 293T cells (5 x 105) were mock transfected (thin line) or transfected with an expression plasmid enabling surface expression of mDR6 (thick line). Cells were incubated on ice for 30 min in 50 μl FACS buffer (PBS, 5% Fetal calf serum, 0.02% azide) containing 1 μg/mL of Luke-1 antibody. After washing in FACS buffer, PE-conjugated antibody to rat IgG was added. Cells were incubated on ice for 30 min, washed and then analyzed by flow cytometry.
DR6 (monoclonal Luke-1) antibody was purified to ≥95% in SDS-PAGE.
PBS containing 0.02% sodium azide
DR6 antibody can be stored at 4°C, stable for one year.
Type:
Antigen: TNFRSF21
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human