Anti-ENO3 Rabbit Polyclonal Antibody
PRSI79-755
- Antigen name:Enolase 3, (beta, muscle)
- Antigen symbol:ENO3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:GSD13|MSE
- Concentration:1 mg/mL
- Immunogen:beta-Enolase (ENO-3) antibody was raised against a peptide sequence around aa.49~53 (L-R-D-G-D) derived from Human β-Enolase (ENO-3).
- Size:100 µg
- Pk:100 µG
Appears to have a function in striated muscle development and regeneration.
Western Blot: 1:500~1:1000
Antibodies were purified by affinity-chromatography using epitope-specific peptide.
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ENO3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat