Anti-CRABP2 Rabbit Polyclonal Antibody
Catalog # PRSI79-748
Supplier: ProSci Inc.
Specifications
- Antigen name:Cellular Retinoic Acid Binding Protein 2
- Antigen symbol:CRABP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:RBP6|CRABP-II
- Concentration:1 mg/mL
- Immunogen:CRABP2 antibody was raised against a peptide sequence around aa.87~91 (K-W-E-S-E) derived from Human CRABP2.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Transports retinoic acid to the nucleus. Regulates the access of retinoic acid to the nuclear retinoic acid receptors
Western Blot: 1:500~1:1000
Antibodies were purified by affinity-chromatography using epitope-specific peptide.
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CRABP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: