Specifications
- Antigen name:Solute Carrier Family 6 (Neurotransmitter Transporter), Member 13
- Antigen symbol:SLC6A13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Rat,Mouse
- Form:Liquid
- Antigen synonyms:GAT-2
- Immunogen:Peptide corresponding to amino acid residues from the C-terminal region of rat GAT-2.
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Gamma-aminobutyric acid (GABA) is the primary inhibitory neurotransmitter in the central nervous system, causing a hyperpolarization of the membrane through the opening of a Cl− channel associated with the GABAA receptor (GABAA-R) subtype. GABA plasma membrane transporters (GATs) influence synaptic neurotransmission by high-affinity uptake and release of GABA. To date, four distinct GABA transporters have been identified: GAT-1, GAT-2, GAT-3, and BGT-1. GAT-2 is found in a wide range of neuronal and non-neuronal cells including dendrites and axon terminals as well as epithelial cells and cells forming the pia and arachnoid complex (Conti et al., 1999).
The antibody has been directly tested for reactivity in immunohistochemistry in rat cortex and retinal sections. It is anticipated that the antibody will also work in mouse sections based on the fact that this species has 100% homology with the amino acid sequence used as antigen.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLC6A13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat