Specifications
- Antigen name:Solute Carrier Family 32 Member A1
- Antigen symbol:SLC32A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Bovine,Dog,Rat
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:VGAT|Viaat
- Immunogen:Peptide corresponding to amino acid residues from the N-terminal region of rat Vesicular GABA Amino Acid Transporter (VGAT).
- Size:100 µl
- Pk:100 µl
Specifications
About this item
The Vesicular GABA Amino Acid Transporter (VGAT) is responsible for transport of the inhibitory neurotransmitter into synaptic vesicles(McIntire et al., 1997). The VGAT protein (also known as the Vesicular Inhibitory Amino Aid Transporter or VIAAT) is expressed in synaptic vesicles of both glycine and GABAergic synapses throughout the CNS (Chaudhry et al., 1998). Expression of the VGAT protein changes during development and also in response to patterns of neuronal activity (De et al., 2005).
Affinity Purified
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLC32A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat, Dog, Bovine