Anti-GPR139 Goat Polyclonal Antibody
Catalog # PRSI46-852
Supplier: ProSci Inc.
Specifications
- Antigen name:G Protein-coupled Receptor 139
- Antigen symbol:GPR139
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Form:Liquid
- Antigen synonyms:PGR3|GPRg1
- Concentration:500 ug/mL
- Immunogen:GPR139 antibody was raised against a 12 amino acid synthetic peptide near the N-Terminus of GPR139.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
GPR139 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
GPR139 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GPR139
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: