Anti-ATF7 Goat Polyclonal Antibody
PRSI45-305
- Antigen name:Activating Transcription Factor 7
- Antigen symbol:ATF7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:ATFA
- Concentration:500 ug/mL
- Immunogen:ATF7 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of ATF7.
- Size:100 µg
- Pk:100 µG
ATF7 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
ATF7 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ATF7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human