Anti-RAB11A Goat Polyclonal Antibody
Catalog # PRSI45-130
Supplier: ProSci Inc.
Specifications
- Antigen name:Ras-related protein Rab-11A
- Antigen symbol:RAB11A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:RAB11A|MGC1490|YL8|Ras-related protein Rab-11A|RAB 11A|member oncogene family|member RAS oncogene family
- Concentration:500 ug/mL
- Immunogen:RAB11A antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of RAB11A.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
RAB11A antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
RAB11A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: RAB11A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse