Order Entry
United Kingdom
ContactUsLinkComponent
Anti-CUFF Goat Polyclonal Antibody
Anti-CUFF Goat Polyclonal Antibody
Catalog # PRSI43-013
Supplier:  ProSci Inc.
undefined
Anti-CUFF Goat Polyclonal Antibody
Catalog # PRSI43-013
Supplier:  ProSci Inc.

Specifications

  • Antigen symbol:
    CUFF
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Form:
    Liquid
  • Concentration:
    500 ug/mL
  • Immunogen:
    cuff antibody was raised against a 14 amino acid peptide near the internal region of cuff (Drosophila melanogaster).
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

cuff antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

cuff antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CUFF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: