Anti-RSL1D1 Goat Polyclonal Antibody
Catalog # PRSI42-504
Supplier: ProSci Inc.
Specifications
- Antigen name:Ribosomal L1 Domain Containing 1
- Antigen symbol:RSL1D1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:RSL1D1|MGC142259|cellular senescence inhibited gene protein|ribosomal L1 domain containing 1|CSIG|L12|DKFZP564M182|PBK1|MGC138433
- Concentration:500 ug/mL
- Immunogen:RSL1D1 antibody was raised against a 14 amino acid synthetic peptide near the internal region of RSL1D1.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
RSL1D1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
RSL1D1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: RSL1D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human