Anti-Crot Goat Polyclonal Antibody
PRSI42-406
- Antigen name:Carnitine O-octanoyltransferase
- Antigen symbol:CROT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Rat
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Carnitine O octanoyltransferase|OCTC_HUMAN|Peroxisomal carnitine acyltransferase|Peroxisomal carnitine O-octanoyltransferase.|Peroxisomal carnitine O octanoyltransferase|CROT|COT
- Concentration:500 ug/mL
- Immunogen:Crot antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of Crot (rat).
- Size:100 µg
- Pk:100 µG
Crot antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
Crot antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: Crot
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: