Anti-DBF4 Goat Polyclonal Antibody
PRSI42-400
- Antigen name:DBF4 Zinc Finger
- Antigen symbol:DBF4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:DBF4A|CHIF|ASK|ZDBF1
- Concentration:500 ug/mL
- Immunogen:Dbf4 antibody was raised against a amino acid synthetic peptide near the of Dbf4 (mouse).
- Size:100 µg
- Pk:100 µG
Dbf4 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
Dbf4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: DBF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat