Anti-MTHFD1L Goat Polyclonal Antibody
PRSI42-396
- Antigen name:Methylenetetrahydrofolate Dehydrogenase (NADP+ dependent) 1-Like
- Antigen symbol:MTHFD1L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Concentration:500 ug/mL
- Immunogen:MTHFD1L antibody was raised against a amino acid synthetic peptide near the of MTHFD1L (aa 535-538).
- Size:100 µg
- Pk:100 µG
MTHFD1L antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
MTHFD1L antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: MTHFD1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human