Order Entry
United Kingdom
ContactUsLinkComponent
Anti-EDC3 Rabbit Polyclonal Antibody
Anti-EDC3 Rabbit Polyclonal Antibody
Catalog # PRSI39-862
Supplier:  ProSci Inc.
undefined
Anti-EDC3 Rabbit Polyclonal Antibody
Catalog # PRSI39-862
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Enhancer of mRNA-decapping protein 3
  • Antigen symbol:
    EDC3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Antigen synonyms:
    YJDC|LSM16|YJEFN2
  • Immunogen:
    A synthetic peptide surrounding the pS161 site.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

EDC3 is associated with an mRNA-decapping complex required for removal of the 5-prime cap from mRNA prior to its degradation from the 5-prime end. It has also been implicated in cell signaling involving the 14-3-3 proteins and related pathways.

Affinity Purified

Type:
Antigen: EDC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat