Anti-YBX3 Rabbit Polyclonal Antibody
PRSI31-225
- Antigen name:Y box binding protein 3
- Antigen symbol:YBX3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:ZONAB|CSDA|CSDA1|DBPA
- Immunogen:A synthetic peptide corresponding a region of human CSDA.
- Size:100 µg
- Pk:100 µG
Human DNA-binding protein (dbpA) is a member of a Y-box binding protein family containing a cold shock domain. The increased expression of Y box binding proteins in somatic cells is associated with cell proliferation and transformation.
Antibody is purified by Protein A/G affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: YBX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human