Anti-TRP53 Rabbit Polyclonal Antibody
PRSI28-625
- Antigen name:Transformation Related Protein 53
- Antigen symbol:TRP53
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:Tp53|bbl|bhy|p44|bfy|p53
- Immunogen:A synthetic peptide corresponding a region of mouse TRP53.
- Size:100 µg
- Pk:100 µG
Trp53 is a protein found in elevated levels in a great variety of transformed cells
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TRP53
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat