Anti-ASIC2 Rabbit Polyclonal Antibody
Catalog # PRSI28-231
Supplier: ProSci Inc.
Specifications
- Antigen name:Acid Sensing Ion Channel Subunit 2
- Antigen symbol:ASIC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Dog
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human ACCN1.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
ACCN1 is a member of the degenerin/epithelial sodium channel (DEG/ENaC) superfamily. ACCN1 may play a role in neurotransmission. In addition, a heteromeric association between ACCN1 and ACCN3 (variant 1) has been observed to co-assemble into proton-gated channels sensitive to gadolinium.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ASIC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human