- Antigen name:LIM Domain Binding 2
- Antigen symbol:LDB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Zebrafish,Human,Dog,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:Ubiquitin specific peptidase 14 (tRNA-guanine transglycosylase)|CLIM1
- Immunogen:A synthetic peptide corresponding a region of human LDB2.
- Size:100 µg
- Pk:100 µG
LDB2 belongs to the LDB family. LDB2 binds to the LIM domain of a wide variety of LIM domain-containing transcription factors. LIM domains are required for both inhibitory effects on LIM homeodomain transcription factors and synergistic transcriptional activation events. The inhibitory actions of the LIM domain can often be overcome by the LIM co-regulators known as CLIM2, LDB2 and NLI. LIM homeoproteins and CLIMs are involved in a variety of developmental processes.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: LDB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog, Zebrafish