- Antigen name:BARX Homeobox 1
- Antigen symbol:BARX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human BARX1.
- Size:50 µg
- Pk:50 µG
BARX1 belongs to the Bar subclass of the homeobox gene family. The function of this gene has not yet been determined; however, studies in the mouse and chick homolog suggest a role in developing teeth and craniofacial mesenchyme of neural crest origin. The role of these homologs implicates the human gene as a candidate for unmapped disorders involving tooth and jaw development.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: BARX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human