Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-ZNF409 Rabbit Polyclonal Antibody
  PRSI27-536
undefined
Anti-ZNF409 Rabbit Polyclonal Antibody
  PRSI27-536

 

  • Antigen name:
    Zinc Finger Protein 409
  • Antigen symbol:
    ZNF409
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Dog
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Immunogen:
    A synthetic peptide corresponding a region of human ZNF409.
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

ZNF209 is a candidate transcription factor

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ZNF409
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human